Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146330.1 | NP_001139802.1 | 1 | 814 |
Set Membership: |
G-protein coupled receptor |