Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077633.1 | NP_001071101.1 | 3, 4 | 210 |
Gene Symbol: Dcpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC024627.1 | - | 191 | 4925312 | ||
BC045522.1 | - | 3, 4 | 201 | 4190591 | ||
BC090399.1 | - | 191 | 4168483 | |||
DQ237936.1 | - | 209 | ||||
S76879.1 | - | 250 | ||||
U03711.1 | - | 214 |
Gene Symbol: Dcpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC059096.1 | - | 2, 3 | 228 | 4914089 | |
BC171996.1 | - | 210 | 9054282 | |||
DQ237937.1 | - | 209 |