Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad54b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039556.3 | NP_001034645.1 | 8 | 1292 | ||
GenBank mRNA | BC057604.1 | - | 8 | 1240 | 6416213 |