Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pramef17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085540.2 | NP_001079009.1 | 1 | 134 |
Gene Symbol: Gm13128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085541.1 | NP_001079010.1 | 1 | 134 |