Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syndig1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033334.2 | NP_001028506.1 | 3 | 881 | ||
GenBank mRNA | AK140195.1 | - | 3 | 881 | ||
AY993933.1 | - | 4 | 930 | |||
AY993934.1 | - | 3 | 905 | |||
BC132221.1 | - | 2 | 598 | 40130498 | ||
BC138501.1 | - | 2 | 598 | 8861521 |