Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp1c-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC141384.1 | - | 8 | 2526 | 9055997 | |
DQ117602.1 | - | 8 | 2530 |