Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198410.3 | NP_940802.1 | 8 | 1566 | ||
GenBank mRNA | AK005086.1 | - | 2 | 805 | ||
AK139144.1 | - | 8 | 1567 | |||
BC030379.1 | - | 3 | 1191 |
Set Membership: |
G-protein coupled receptor |