Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006932.2 | NP_001006933.2 | 20 | 2332 | ||
NM_001318936.1 | NP_001305865.1 | 21 | 2383 | |||
NM_001318937.1 | NP_001305866.1 | 17 | 2040 | |||
NM_001318938.1 | NP_001305867.1 | 19 | 1919 | |||
NM_021135.5 | NP_066958.2 | 19 | 2185 | |||
XM_006715549.3 | XP_006715612.1 | 20 | 2214 | |||
GenBank mRNA | AB209116.1 | - | 21 | 2146 | ||
AK095751.1 | - | 20 | 2293 | |||
AK295674.1 | - | 19 | 1993 | |||
AK307470.1 | - | 4 | 455 | |||
AK310428.1 | - | 19 | 1912 | |||
BC002363.2 | - | 21 | 2145 | 2961482 | ||
KJ534936.1 | - | 5 | 469 | |||
KJ535080.1 | - | 17 | 1659 | |||
KJ535087.1 | - | 20 | 1950 | |||
X85106.1 | - | 21 | 2101 |