Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007951.3 | NP_031977.1 | 4 | 337 | ||
GenBank mRNA | AK010451.1 | - | 4 | 282 | ||
AK080617.1 | - | 4 | 310 | |||
AK168188.1 | - | 4 | 291 | |||
AK171834.1 | - | 4 | 283 | |||
BC083141.1 | - | 4 | 272 | 30294607 | ||
BC099476.1 | - | 4 | 270 | 30847983 | ||
BC107315.1 | - | 4 | 261 | 40053362 | ||
BC107316.1 | - | 4 | 261 | 40053363 | ||
BC132421.1 | - | 4 | 265 | 40130698 | ||
BC132423.1 | - | 4 | 265 | 40130700 | ||
BY085685.1 | - | 4 | 336 | |||
D73368.1 | - | 4 | 283 | |||
U66870.1 | - | 4 | 266 |