Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007917.3 | NP_031943.3 | 7 | 612 | ||
GenBank mRNA | AK146757.1 | - | 6 | 492 | ||
AK167097.1 | - | 6 | 501 | |||
BC010759.1 | - | 6 | 486 | 4216609 | ||
BC085087.1 | - | 6 | 484 | 6438419 | ||
M61731.1 | - | 6 | 498 |