Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010917.2 | NP_035047.2 | 19 | 3630 | ||
GenBank mRNA | AK084876.1 | - | 19 | 3569 | ||
AK144878.1 | - | 19 | 3595 | |||
AK166779.1 | - | 19 | 3566 | |||
BC052939.1 | - | 19 | 3625 | |||
BC131668.1 | - | 6 | 806 | |||
BC131669.1 | - | 19 | 3541 | 40110859 | ||
X14194.1 | - | 19 | 3541 | |||
X14480.1 | - | 19 | 3551 |