Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mplkip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025479.5 | NP_079755.1 | 2 | 537 | ||
GenBank mRNA | AK011919.1 | - | 2 | 532 | ||
AK012770.1 | - | 2 | 530 | |||
AK030630.1 | - | 2 | 538 | |||
AK141333.1 | - | 1540 | ||||
BC139338.1 | - | 2 | 490 | 8862360 | ||
BC139341.1 | - | 2 | 490 | 8862363 |
Gene Symbol: Gm7102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177513.1 | NP_001170984.1 | 1 | 779 | ||
GenBank mRNA | AK163227.1 | - | 1 | 783 |