Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aff3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290814.1 | NP_001277743.1 | 19 | 3339 | ||
GenBank mRNA | BC052061.1 | - | 19 | 2669 | ||
BQ443842.1 | - | 3 | 179 |