Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp26c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105201.1 | NP_001098671.1 | 5 | 1053 | ||
GenBank mRNA | BC151098.1 | - | 5 | 1152 | 9088011 | |
BC151106.1 | - | 5 | 1152 | 9088019 |