Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206537.2 | NP_996260.1 | 4, 5 | 649 | ||
GenBank mRNA | AI047349.1 | - | 1, 2 | 85 | ||
AI181879.1 | - | 4, 5 | 638 | |||
BC043929.1 | - | 3, 4 | 431 |
Gene Symbol: Cyp2c50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134144.2 | NP_598905.2 | 4, 5 | 668 | ||
GenBank mRNA | AY206873.1 | - | 4, 5 | 647 | ||
BC011222.1 | - | 4, 5 | 651 | 4195223 |