Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcnka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146307.1 | NP_001139779.1 | 15 | 1707 | ||
NM_024412.3 | NP_077723.3 | 16 | 1711 | |||
GenBank mRNA | AY373832.1 | - | 15 | 1662 |