Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SALL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291446.1 | NP_001278375.1 | 3 | 1445 | ||
NM_001291447.1 | NP_001278376.1 | 3 | 1425 | |||
NM_005407.2 | NP_005398.2 | 2 | 1850 | |||
XM_011537064.1 | XP_011535366.1 | 2 | 1776 | |||
XM_011537065.1 | XP_011535367.1 | 2 | 1922 | |||
GenBank mRNA | AB002358.2 | - | 2 | 1837 | ||
AK293574.1 | - | 3 | 1131 | |||
AK296414.1 | - | 3 | 1244 | |||
AK299931.1 | - | 2 | 1724 | |||
BC136528.1 | - | 2 | 1585 | 9020518 | ||
BC136529.1 | - | 2 | 1585 | 9020519 | ||
X98834.1 | - | 2 | 1737 |