Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010120.5 | NP_034250.3 | 3 | 624 | ||
GenBank mRNA | AF026481.1 | - | 2 | 1443 | ||
AK009026.1 | - | 3 | 595 | |||
AK013268.1 | - | 3 | 623 | |||
AK019451.1 | - | 3 | 625 | |||
AK150651.1 | - | 3 | 621 | |||
AK150673.1 | - | 3 | 597 | |||
AK150677.1 | - | 3 | 597 | |||
AK166368.1 | - | 596 | ||||
AK167333.1 | - | 3 | 584 | |||
BC027437.1 | - | 3 | 586 | 4017054 | ||
CT010335.1 | - | 1 | 271 |
Gene Symbol: Gm2016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122662.1 | NP_001116134.1 | 2 | 762 | ||
GenBank mRNA | AK163212.1 | - | 2 | 762 |
Gene Symbol: Gm2022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177574.1 | NP_001171045.1 | 2 | 740 | ||
GenBank mRNA | AK146348.1 | - | 637 |
Gene Symbol: Gm21319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270722.1 | NP_001257651.1 | 2 | 771 | ||
GenBank mRNA | AK139496.1 | - | 2 | 773 |
Gene Symbol: Gm5039 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC094938.1 | - | 1 | 552 |
Gene Symbol: Gm8300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177565.1 | NP_001171036.1 | 2 | 774 | ||
GenBank mRNA | AK139584.1 | - | 2 | 774 |