Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vipr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009511.2 | NP_033537.1 | 8 | 905 | ||
GenBank mRNA | AK038743.1 | - | 8 | 906 | ||
BC138572.1 | - | 8 | 803 | 8861592 | ||
D28132.1 | - | 8 | 837 |
Set Membership: |
G-protein coupled receptor |