Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002970.3 | NP_002961.1 | 4 | 487 | ||
NR_027783.2 | 5 | 597 | ||||
XM_017029720.1 | XP_016885209.1 | 1 | 1115 | |||
GenBank mRNA | AK312162.1 | - | 4 | 470 | ||
AL050290.1 | - | 5 | 558 | |||
AY841998.1 | - | 5 | 404 | |||
BC002503.2 | - | 4 | 448 | 3051095 | ||
BC008424.1 | - | 4 | 472 | 4134501 | ||
BT006825.1 | - | 4 | 293 | |||
CD516700.1 | - | 4 | 493 | |||
CD638929.1 | - | 5 | 585 | |||
CR450294.1 | - | 4 | 293 | |||
M55580.1 | - | 298 | ||||
M77693.1 | - | 4 | 458 |