Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001036293.2 | NP_001031370.1 | 4 | 649 | ||
GenBank mRNA | AK002810.1 | - | 4 | 651 | ||
AK004200.1 | - | 4 | 571 | |||
AK208090.1 | - | 1 | 71 | |||
BC019448.1 | - | 4 | 649 | 4212132 |