Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102565.1 | NP_001096035.1 | 1 | 148 | ||
GenBank mRNA | AK042102.1 | - | 1 | 149 | ||
AK143105.1 | - | 1 | 146 | |||
AK144828.1 | - | 1 | 149 | |||
BC050856.1 | - | 1 | 135 | |||
BC094381.1 | - | 1 | 137 |
Gene Symbol: Nrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013372.2 | NP_001013390.1 | 1 | 326 | ||
GenBank mRNA | AK012518.1 | - | 1 | 145 | ||
AK049549.1 | - | 1 | 149 | |||
AY753183.1 | - | 1 | 326 | |||
BC119399.1 | - | 1 | 303 | 8734148 | ||
BC119401.1 | - | 1 | 303 | 8734150 |