Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdr39u1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082975.1 | NP_001076444.1 | 6 | 629 | ||
GenBank mRNA | AK009339.1 | - | 6 | 609 | ||
AK049709.1 | - | 6 | 623 | |||
BC031783.1 | - | 1 | 951 | |||
BC058857.1 | - | 4 | 893 | |||
BC087941.1 | - | 6 | 608 | 30303350 |