Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000332.3 | NP_000323.2 | 8 | 1378 | ||
NM_001128164.1 | NP_001121636.1 | 7 | 1329 | |||
GenBank mRNA | BC117125.1 | - | 1 | 440 | 40125676 | |
JX901140.1 | - | 1 | 378 | |||
X79204.1 | - | 8 | 1342 |