Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nova1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021361.1 | NP_067336.1 | 2 | 285 | ||
GenBank mRNA | AF232828.1 | - | 2 | 219 | ||
AK090295.1 | - | 2 | 574 | |||
BM942596.1 | - | 2 | 285 |