Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crxos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033638.2 | NP_001028810.1 | 4, 5 | 620 | ||
NM_001145190.2 | NP_001138662.1 | 2, 3 | 257 | |||
GenBank mRNA | AB472692.1 | - | 4, 5 | 573 | ||
AB472694.1 | - | 2, 3 | 241 | |||
AK145701.1 | - | 4, 5 | 620 | |||
AY590890.1 | - | 2, 3 | 314 | |||
AY590891.1 | - | 3, 4 | 488 | |||
CJ040988.1 | - | 2, 3 | 240 |
Gene Symbol: Gm9376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101609.1 | NP_001095079.1 | 1 | 301 |