Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145985.4 | NP_666097.3 | 10 | 1709 | ||
GenBank mRNA | AK028556.1 | - | 10 | 1714 | ||
AK047995.1 | - | 10 | 1749 | |||
AK128973.1 | - | 10 | 1714 | |||
AK147747.1 | - | 10 | 1743 | |||
AK154066.1 | - | 1713 | ||||
AK170005.1 | - | 6 | 896 | |||
BC017124.1 | - | 10 | 1704 | 4480874 | ||
BC023728.1 | - | 10 | 1728 | 5351777 | ||
BC024127.1 | - | 9 | 1329 | |||
BC033387.1 | - | 10 | 1709 | 5368060 | ||
BC034754.1 | - | 10 | 1686 | 4954556 | ||
BC083152.1 | - | 1702 | 30461105 |