Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302123.1 | NP_001289052.1 | 2 | 371 | ||
NM_005409.4 | NP_005400.1 | 2 | 371 | |||
GenBank mRNA | AF002985.1 | - | 2 | 248 | ||
AF030514.1 | - | 2 | 230 | |||
AF352781.1 | - | 2 | 239 | |||
AL703031.1 | - | 2 | 211 | |||
BC005292.1 | - | 2 | 221 | 3934139 | ||
BC012532.1 | - | 2 | 375 | 4096572 | ||
BC110986.1 | - | 2 | 230 | 5178174 | ||
BP289021.1 | - | 2 | 273 | |||
BT006787.1 | - | 2 | 161 | |||
U59286.1 | - | 2 | 183 | |||
U66096.1 | - | 2 | 206 | |||
Y15220.1 | - | 2 | 167 |