Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdpoz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148949.2 | NP_683751.2 | 2 | 2056 | ||
GenBank mRNA | BB709516.1 | - | 1 | 190 |
Gene Symbol: Gm9125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163730.2 | NP_001157202.1 | 2 | 2089 | ||
GenBank mRNA | AK033449.1 | - | 2 | 2090 |