Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000609.6 | NP_000600.1 | 3 | 300 | ||
NM_001033886.2 | NP_001029058.1 | 3 | 300 | |||
NM_001178134.1 | NP_001171605.1 | 3 | 300 | |||
NM_199168.3 | NP_954637.1 | 3 | 300 | |||
GenBank mRNA | AK292628.1 | - | 3 | 303 | ||
AK311814.1 | - | 3 | 298 | |||
AY644456.1 | - | 3 | 257 | |||
AY874118.1 | - | 3 | 287 | |||
BC031072.1 | - | 3 | 292 | |||
BC039893.1 | - | 3 | 278 | 5729604 | ||
CD172464.1 | - | 3 | 298 | |||
CR450283.1 | - | 3 | 208 | |||
DA266752.1 | - | 3 | 301 | |||
DQ345517.1 | - | 3 | 252 | |||
DQ345518.1 | - | 3 | 247 | |||
DQ345519.1 | - | 3 | 252 | |||
DQ345520.1 | - | 3 | 252 | |||
L36033.1 | - | 3 | 287 | |||
L36034.1 | - | 3 | 287 | |||
U16752.1 | - | 3 | 288 |