Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293791.1 | NP_001280720.1 | 18 | 3215 | ||
NM_001293792.1 | NP_001280721.1 | 18 | 3215 | |||
NM_027590.4 | NP_081866.1 | 17 | 3137 | |||
GenBank mRNA | AK030974.1 | - | 17 | 3100 | ||
AK034498.1 | - | 18 | 3132 | |||
AK147743.1 | - | 18 | 3190 |