Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039677.2 | NP_001034766.1 | 5 | 959 | ||
GenBank mRNA | AK031425.1 | - | 5 | 755 | ||
BC013478.1 | - | 4 | 979 | |||
BC113751.1 | - | 5 | 959 | 40057211 | ||
BC147704.1 | - | 5 | 790 | 9087463 | ||
BC147707.1 | - | 5 | 790 | 9087466 |