Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4bpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016995.2 | NP_058691.2 | 6 | 996 | ||
XM_006249705.2 | XP_006249767.1 | 7 | 1052 | |||
XM_008769423.1 | XP_008767645.1 | 7 | 869 | |||
GenBank mRNA | BC088152.1 | - | 6 | 996 | 7322205 | |
FQ209636.1 | - | 6 | 1007 | |||
FQ218474.1 | - | 6 | 977 | |||
Z50052.1 | - | 6 | 975 |