Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093770.2 | NP_001087239.2 | 5 | 468 | ||
NM_001164644.1 | NP_001158116.1 | 5 | 438 | |||
NM_001164645.1 | NP_001158117.1 | 5 | 324 | |||
NM_001164646.1 | NP_001158118.1 | 5 | 332 | |||
NM_001164647.1 | NP_001158119.1 | 4 | 408 | |||
NM_005411.4 | NP_005402.3 | 5 | 479 | |||
XM_005270062.4 | XP_005270119.1 | 4 | 468 | |||
XM_006717953.2 | XP_006718016.1 | 5 | 461 | |||
GenBank mRNA | AK290703.1 | - | 5 | 470 | ||
AK298002.1 | - | 5 | 324 | |||
AK298029.1 | - | 4 | 357 | |||
AK298034.1 | - | 4 | 215 | |||
AK298092.1 | - | 3 | 210 | |||
AK309518.1 | - | 4 | 480 | |||
AK316196.1 | - | 4 | 132 | |||
BC029913.2 | - | 4 | 435 | |||
BC111570.1 | - | 4 | 387 | 40069324 | ||
BC171875.1 | - | 4 | 398 | 9054529 | ||
BI820937.1 | - | 5 | 462 | |||
DA588383.1 | - | 5 | 332 | |||
HQ021433.2 | - | 4 | 408 | |||
HQ021434.1 | - | 4 | 408 | |||
HQ021435.1 | - | 4 | 408 | |||
HQ021436.1 | - | 4 | 408 | |||
HQ021437.1 | - | 5 | 479 | |||
HQ021438.1 | - | 5 | 479 | |||
HQ021439.1 | - | 5 | 479 | |||
HQ021440.1 | - | 5 | 468 | |||
HQ021441.1 | - | 5 | 468 | |||
HQ021442.1 | - | 5 | 468 | |||
JX502764.1 | - | 5 | 479 | |||
JX502765.1 | - | 5 | 468 |