Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012522.2 | NP_036654.2 | 14 | 1579 | ||
GenBank mRNA | D01098.1 | - | 14 | 1580 | ||
FQ211441.1 | - | 14 | 1638 | |||
M88344.1 | - | 14 | 1728 | |||
M88345.1 | - | 12 | 2108 | |||
M88346.1 | - | 13 | 1474 | |||
M88347.1 | - | 14 | 1590 |