Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000698.4 | NP_000689.1 | 4 | 626 | ||
NM_001256153.2 | NP_001243082.1 | 4 | 626 | |||
NM_001256154.2 | NP_001243083.1 | 4 | 626 | |||
NM_001320861.1 | NP_001307790.1 | 4 | 626 | |||
NM_001320862.1 | NP_001307791.1 | 4 | 664 | |||
XM_017016012.1 | XP_016871501.1 | 3 | 501 | |||
GenBank mRNA | AB208946.1 | - | 5 | 2897 | ||
AK225603.1 | - | 4 | 579 | |||
AK315223.1 | - | 4 | 576 | |||
BC034464.1 | - | 4 | 565 | |||
BC040450.1 | - | 4 | 555 | |||
BC130332.1 | - | 4 | 579 | 40146363 | ||
BC132677.1 | - | 4 | 579 | 40146699 | ||
BC143985.1 | - | 4 | 579 | 9052503 | ||
HM592258.1 | - | 4 | 512 | |||
HM592259.1 | - | 4 | 512 | |||
HM592260.1 | - | 4 | 512 | |||
HM592261.1 | - | 4 | 512 | |||
J03571.1 | - | 4 | 546 | |||
J03600.1 | - | 4 | 556 | |||
JQ728550.1 | - | 3 | 432 |