Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012582.2 | NP_036714.2 | 4 | 222 | ||
GenBank mRNA | AF476963.1 | - | 4 | 253 | ||
AY325231.1 | - | 4 | 248 | |||
BC089816.1 | - | 4 | 222 | 7321960 | ||
FQ209575.1 | - | 2 | 127 | |||
FQ209592.1 | - | 4 | 274 | |||
FQ209684.1 | - | 4 | 274 | |||
FQ211144.1 | - | 4 | 276 | |||
FQ219122.1 | - | 4 | 274 | |||
FQ219413.1 | - | 1 | 32 | |||
FQ219443.1 | - | 4 | 274 | |||
FQ219547.1 | - | 4 | 245 | |||
FQ219598.1 | - | 4 | 244 |