Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077641.1 | NP_001071109.1 | 2, 3 | 312 | ||
XM_008762247.1 | XP_008760469.1 | 1, 2 | 459 | |||
GenBank mRNA | L14323.2 | - | 1, 2 | 568 | ||
M20636.1 | - | 2, 3 | 312 |