Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006610.1 | NP_001006611.1 | 2 | 1168 | ||
NM_003031.3 | NP_003022.3 | 2 | 796 | |||
XM_006721246.1 | XP_006721309.1 | 3 | 972 | |||
XM_011523279.1 | XP_011521581.1 | 6 | 2312 | |||
GenBank mRNA | AB451253.1 | - | 1 | 679 | ||
AB451377.1 | - | 1 | 679 | |||
AK094663.1 | - | 1 | 2133 | |||
AK225895.1 | - | 2 | 829 | |||
AK313861.1 | - | 2 | 1182 | |||
BC035562.1 | - | 2 | 796 | 5224755 | ||
BC042550.1 | - | 2 | 1138 | 5198176 | ||
BC044920.1 | - | 6 | 2275 | 5295925 | ||
BX647064.1 | - | 1 | 1740 | |||
U63295.1 | - | 1 | 679 | |||
U76247.1 | - | 2 | 865 |