Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052800.1 | NP_434687.1 | 3, 4 | 279 | ||
GenBank mRNA | AW919734.1 | - | 1, 2 | 116 | ||
CV072953.1 | - | 3, 4 | 278 | |||
FQ229684.1 | - | 3, 4 | 319 | |||
FQ234584.1 | - | 289 | ||||
FQ234769.1 | - | 289 | ||||
M22642.1 | - | 3, 4 | 276 |