Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017060.2 | NP_058756.2 | 3 | 161 | ||
XM_006230687.2 | XP_006230749.1 | 2 | 1030 | |||
GenBank mRNA | AB298805.1 | - | 1 | 80 | ||
CB613004.1 | - | 3 | 161 | |||
FQ224126.1 | - | 3 | 221 |