Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109892.1 | NP_001103362.1 | 14 | 2543 | ||
NM_001109893.1 | NP_001103363.1 | 13 | 2279 | |||
NM_001109894.1 | NP_001103364.1 | 13 | 2276 | |||
NM_001109895.1 | NP_001103365.1 | 12 | 2201 | |||
NM_001109896.1 | NP_001103366.1 | 12 | 2198 | |||
NM_012712.1 | NP_036844.1 | 14 | 2546 | |||
XM_006230389.2 | XP_006230451.1 | 14 | 2541 | |||
XM_006230390.2 | XP_006230452.1 | 14 | 2538 | |||
XM_006230392.2 | XP_006230454.1 | 14 | 2547 | |||
XM_006230393.2 | XP_006230455.1 | 12 | 2193 | |||
XM_008759861.1 | XP_008758083.1 | 13 | 4703 | |||
GenBank mRNA | CB780941.1 | - | 2 | 117 | ||
L19109.1 | - | 6 | 680 | |||
L19110.1 | - | 6 | 684 | |||
U59800.1 | - | 2 | 223 | |||
Z35138.1 | - | 11 | 1549 | |||
Z35139.1 | - | 10 | 1447 |