Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109892.1 | NP_001103362.1 | 12 | 2268 | ||
NM_001109893.1 | NP_001103363.1 | 11 | 2004 | |||
NM_001109894.1 | NP_001103364.1 | 11 | 2001 | |||
NM_001109895.1 | NP_001103365.1 | 10 | 1926 | |||
NM_001109896.1 | NP_001103366.1 | 10 | 1923 | |||
NM_012712.1 | NP_036844.1 | 12 | 2271 | |||
XM_006230389.2 | XP_006230451.1 | 12 | 2266 | |||
XM_006230390.2 | XP_006230452.1 | 12 | 2263 | |||
XM_006230392.2 | XP_006230454.1 | 12 | 2272 | |||
XM_006230393.2 | XP_006230455.1 | 10 | 1918 | |||
XM_008759861.1 | XP_008758083.1 | 11 | 4428 | |||
GenBank mRNA | L19109.1 | - | 4 | 405 | ||
L19110.1 | - | 4 | 409 | |||
Z35138.1 | - | 9 | 1274 | |||
Z35139.1 | - | 8 | 1172 |