siRNA Details
Gene Symbol: Onecut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022671.2 | NP_073162.1 | 1 | 677 | ||
XM_006243404.2 | XP_006243466.1 | 1 | 847 | |||
GenBank mRNA | Y14933.1 | - | 1 | 468 |

More Information
Other Species Targeted

Panther Classification:
