Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012777.1 | NP_036909.1 | 2, 3 | 187 | ||
GenBank mRNA | FQ212999.1 | - | 2, 3 | 187 | ||
X55572.1 | - | 2, 3 | 187 |