Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012801.1 | NP_036933.1 | 6 | 790 | ||
XM_006248918.2 | XP_006248980.1 | 4 | 772 | |||
XM_006248919.2 | XP_006248981.1 | 4 | 772 | |||
XM_008768940.1 | XP_008767162.1 | 5 | 2371 | |||
GenBank mRNA | BC061731.1 | - | 4 | 777 | 5598667 | |
D10106.1 | - | 4 | 348 | |||
L06238.1 | - | 6 | 790 | |||
L06894.1 | - | 5 | 434 | |||
S57864.1 | - | 2 | 135 | |||
Z14120.1 | - | 4 | 284 |