Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012817.1 | NP_036949.1 | 3 | 1216 | ||
GenBank mRNA | BC087030.1 | - | 1418 | |||
M62781.1 | - | 3 | 1216 |