Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103692785 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770106.1 | XP_008768328.1 | 1 | 123 |
Gene Symbol: LOC103690821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773208.1 | XP_008771430.1 | 1 | 89 |
Gene Symbol: LOC103692831 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765320.1 | XP_008763542.1 | 1 | 119 |
Gene Symbol: LOC103693375 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770613.1 | XP_008768835.1 | 1 | 122 |
Gene Symbol: LOC103694169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771991.1 | XP_008770213.1 | 1 | 119 |
Gene Symbol: LOC103694404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772938.1 | XP_008771160.1 | 1 | 124 |
Gene Symbol: Rpl39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012875.2 | NP_037007.1 | 89 | |||
GenBank mRNA | BC058489.1 | - | 123 | 6887843 | ||
X82551.1 | - | 89 |