Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012884.1 | NP_037016.1 | 21 | 3011 | ||
XM_006249746.2 | XP_006249808.1 | 22 | 3225 | |||
XM_008769427.1 | XP_008767649.1 | 21 | 3111 | |||
GenBank mRNA | M31725.1 | - | 21 | 3011 |
Set Membership: |
G-protein coupled receptor |