Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003041.3 | NP_003032.1 | 10 | 1229 | ||
XM_006721072.3 | XP_006721135.2 | 10 | 1812 | |||
GenBank mRNA | AK054830.1 | - | 2 | 474 | ||
M95549.1 | - | 10 | 1230 |